missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain AstiPure™ RF2 Series PFA Male Elbow Tube Connector
Brand: Saint-Gobain RF20413
Additional Details : Weight : 0.80000kg
Specifications
PFA | |
Male Elbow Tube Connector | |
1 Each | |
1/4 in. NPT |
38 mm | |
For 4 x 6mm dia. tubing | |
13mm | |
31 mm |