Learn More
Sartorius™ Ash-free Quantitative Filter Papers, Grade 393
Used for quantitative and gravimetric analyses as well as for pressure or vacuum filtration
Brand:  Sartorius™ FT-2-127-580580
Product Code. 10026861
  • 745.00€ / Pack of 100
Estimated Shipment: 22-04-2025
Description
  • Made of 100% cotton linters
  • Ash-free (ash content ≤0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sheet
1 to 2 μm
Suitable for quantitative analysis, Buchner funnels, pressure or vacuum filtration
100 Pk.
Quantitative Filter Paper
580 mm
Cotton Linters, Refined Pulp
300 μm/sec.
393
0.18 mm
580 x 580 mm
580 mm