Learn More
Thermo Scientific™ Arctic Express™ Cryogenic Shippers with Datalogger
Maintain the cold chain and your peace of mind with Thermo Scientific™ Arctic Express™ cryogenic shippers with datalogger.
POA
Specifications
Temperature Range -150°C
Certifications/Compliance 21CFR11 (Datalogger)
Includes Padlock, shipping shell, datalogger
View More Specs
Alert:
NOTE: Not available outside North America
Description
Description
Thermo Scientific™ Arctic Express™ cryogenic shippers are designed to ensure the maintenance of sample integrity when movement of your samples is required. Arctic Express shippers come available with a built-in 21CFR, part 11 compliant datalogger, providing you with additional peace of mind that your important samples have been maintained during transport.
Not available outside North America.
  • Innovative material absorbs liquid nitrogen to prevent spillage during shipment
  • 14- and 21-day liquid nitrogen holding times allow specimens to be shipped safely
  • Durable construction and sturdy base allow dry shipper to withstand rough handling
  • Lockable lid precents unauthorized entry
  • Easily transported via most common carriers
  • Lightweight aluminum design and convenient pail-style handle
  • Warranty varies by country, please contact us for details
  • Specifications
    Specifications
    -150°C
    21CFR11 (Datalogger)
    Padlock, shipping shell, datalogger