Learn More
Thermo Scientific™ Arctic Express™ Cryogenic Shippers with Datalogger
Maintain the cold chain and your peace of mind with Thermo Scientific™ Arctic Express™ cryogenic shippers with datalogger.
POA
Specifications
Temperature Range | -150°C |
---|---|
Certifications/Compliance | 21CFR11 (Datalogger) |
Includes | Padlock, shipping shell, datalogger |
Alert:
NOTE: Not available outside North America
Description
Thermo Scientific™ Arctic Express™ cryogenic shippers are designed to ensure the maintenance of sample integrity when movement of your samples is required. Arctic Express shippers come available with a built-in 21CFR, part 11 compliant datalogger, providing you with additional peace of mind that your important samples have been maintained during transport.
Not available outside North America.
- Innovative material absorbs liquid nitrogen to prevent spillage during shipment
- 14- and 21-day liquid nitrogen holding times allow specimens to be shipped safely
- Durable construction and sturdy base allow dry shipper to withstand rough handling
- Lockable lid precents unauthorized entry
- Easily transported via most common carriers
- Lightweight aluminum design and convenient pail-style handle
Specifications
-150°C | |
21CFR11 (Datalogger) | |
Padlock, shipping shell, datalogger |