missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ APOC3 (Human) Recombinant Protein
Human APOC3 full-length ORF (AAH27977.1, 1 a.a. - 99 a.a.) recombinant protein with GST tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH27977.1 |
---|---|
Gene ID (Entrez) | 345 |
Name | apolipoprotein C-III |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16121721
|
Abnova™
H00000345-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16131721
|
Abnova™
H00000345-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
- Sequence: MQPRVLLVVALLALLASARASEAEDASLLSFMQGYMKHATKTAKDALSSVQESQVAQQARGWVTDGFSSLKDYWSTVKDKFSEFWDLDPEVRPTSAVAA
Specifications
AAH27977.1 | |
apolipoprotein C-III | |
125% SDS-PAGE Stained with Coomassie Blue | |
APOCIII, MGC150353 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
345 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APOC3 | |
GST |