Learn More
Vα2 TCR Rat anti-Mouse, FITC, Clone: B20.1, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553288
Additional Details : Weight : 0.01000kg
Description
The B20.1 antibody reacts with most members of the Vα2 T-cell Receptor (TCR) subfamily in mice having the a, b, and c haplotypes of the Tcrb gene complex. B20.1 antibody may cross-react with Vδ8 TCR, which shares >90% sequence homology with
Host Species: Rat
Clone: B20.1
Isotype: IgG2a λ
Species Reactivity: Mouse
Immunogen: Soluble αβ TCR from mouse cytotoxic T-cell clone KB5-C20
Flow Cytometry
Specifications
Vα2 TCR | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Soluble αβ TCR from mouse cytotoxic T-cell clone KB5-C20 | |
0.25 mg | |
Immunology | |
Mouse | |
IgG2a λ |
Flow Cytometry | |
B20.1 | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.