missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGGT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543360
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AAVRIVPEWQ DYDQEIKQLQ IRFQKEKETG ALYKEKTKEP SREGPQKREE Sequence homology: Human: 100%; Pig: 100%
UGGT1 Polyclonal antibody specifically detects UGGT1 in Human samples. It is validated for Western Blot.Specifications
UGGT1 | |
Polyclonal | |
Unconjugated | |
Uggt1 | |
0910001L17Rik; A930007H10Rik; AA589501; AI414429; AI448372; C820010P03Rik; Gt; HUGT1; RUGT; rUGT1; UDP--Glc:glycoprotein glucosyltransferase; UDP-glucose ceramide glucosyltransferase-like 1; UDP-glucose glycoprotein glucosyltransferase 1; UDP-glucose glycoprotein: glucosyltransferase UGGT; UDP-glucose:glycoprotein glucosyltransferase; UDP-glucose:glycoprotein glucosyltransferase 1; UGCGL1; Uggt; Uggt1; Ugt1; Ugtr | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56886 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NYU2 | |
Uggt1 | |
Synthetic peptide directed towards the middle region of human UGCGL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |