Learn More
thioredoxin, Mouse, Clone: 6C10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TXN.
Brand: Abnova H00007295-M04.100ug
Additional Details : Weight : 0.00010kg
Description
Thioredoxin is a 12kDa oxidoreductase enzyme containing a dithiol-disulfide active site. It is ubiquitous and found in many organisms from plants and bacteria to mammals. Multiple in vitro substrates for thioredoxin have been identified, including ribonuclease, choriogonadotropins, coagulation factors, glucocorticoid receptor, and insulin. Reduction of insulin is classically used as an activity test.[supplied by OMIM
Sequence: MVKQIESKTAFQEALDAAGDKLVVVDFSATWCGPCKMIKPFFHSLSEKYSNVIFLEVDVDDCQDVASECEVKCMPTFQFFKKGQKVGEFSGANKEKLEATINELVSpecifications
thioredoxin | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC003377 | |
TXN | |
TXN (AAH03377, 1 a.a. ∼ 105 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
7295 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Immunofluorescence, Western Blot | |
6C10 | |
Mouse monoclonal antibody raised against a partial recombinant TXN. | |
TXN | |
DKFZp686B1993/MGC61975/TRX/TRX1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |