Learn More
Syntaxin 8 Mouse, Unlabeled, Clone: 48, BD
Mouse Monoclonal Antibody
321.00€ - 634.00€
Specifications
Antigen | Syntaxin 8 |
---|---|
Clone | 48 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15805899
|
BD Biosciences
611352 |
50 μg |
321.00€
1 set |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15815899
|
BD Biosciences
611353 |
150 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
Eukaryotic protein trafficking involves the packaging of molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their components into an acceptor compartment. Recognition between vesicle and acceptor membrane is mediated by the pairing of the integral membrane SNARE proteins. The stable interaction between vesicle proteins (v-SNAREs) and target proteins (t-SNAREs) juxtaposes the membranes and results in an activated docking state and/or membrane fusion. The syntaxin protein family contains a number of members that serve as functional t-SNAREs. One member of this family, syntaxin 8, has two coiled-coil domains, one in the N-terminus and one toward the C-terminus. The C-terminal domain is homologous to a similar domain in syntaxin 6 and is predicted to interact in the formation of SNARE complexes, while a hydrophobic domain in the C-terminus may be involved in membrane anchoring. Syntaxin-8 is ubiquitously expressed, but is present at high levels in the heart and localizes specifically to the ER. Thus, syntaxin-8 is a t-SNARE that is thought to be involved in the early secretory pathway of many different cell types.
Immunofluorescence, Western Blotting
Specifications
Syntaxin 8 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a | |
Affinity Purified |
48 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Mouse Syntaxin 8 aa. 57-164 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.