Learn More
Syntaxin 6 Mouse, Unlabeled, Clone: 30, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610635
Additional Details : Weight : 0.01000kg
Description
Signal transmission between cells is often regulated by the secretion of neurotransmitters or hormones into the extracellular space. This process is controlled by a complex pathway of membrane trafficking which leads to membrane fusion and secretion. Syntaxin 1 is involved in three important complexes that modulate the secretion process: syntaxin and n-sec-1, syntaxin, VAMP, and SNAP-25, and syntaxin, VAMP, SNAP-25, αSNAP, and NSF (20S complex). Syntaxin 6 is a 255 amino acid member of the syntaxin family. It contains a C-terminal transmembrane domain and is located at the Golgi apparatus. In addition, Syntaxin 6 displays significant (47% identity) homology with SNAP-25. In vitro, Syntaxin 6 has been reported to interact with α-SNAP. Syntaxin 6 has been reported to have a calculated molecular weight of 30.6kDa, but has been observed to migrate in a range inbetween ∽ 31-35kDa.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Syntaxin 6 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Canine, Chicken, Frog, Human, Mouse, Rat | |
IgG1 |
Western Blot | |
30 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.