Learn More
Syk w/control Mouse anti-Human, Unlabeled, Clone: 4D10, BD
Mouse Monoclonal Antibody with Control
452.00€
Specifications
Antigen | Syk w/control |
---|---|
Clone | 4D10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15826708
|
BD Biosciences
551816 |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
Syk is a member of the family of non-receptor type protein-tyrosine kinases and plays a crucial role in lymphocyte signaling and development. Syk is expressed in all haematopoietic cells and contributes to the signal transduction process by binding to a tyrosine-based activation motif (ITAM) of immune receptors, including Igα, TCRζ, CD3ε, FcεRIβ, and FceRIγ. Upon receptor activation, Syk binds to phosphorylated ITAMs via its two N-terminal SH2 domains thereby activating Syk and causing tyrosines in Syk to undergo auto-phosphorylation or phosphorylation. These phosphorylated sites can act as binding sites for other signaling molecules or help to regulate enzymatic activity. For example, in mast cells, Syk can activate downstream targets such as phospholipase Cγ1 and VAV. Studies in Syk-/- mast cells identified defects in both the ERK-MAP and JNK-MAP kinase pathways. Syk migrates as 72kDa in SDS/PAGE. (Predicted MW from SWISS-PROT is 72kDa; Accession number P43405).
Host Species: Mouse
Clone: 4D10
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Human Syk peptide aa. 314-339
Western Blotting
Specifications
Syk w/control | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Human Syk peptide aa. 314-339 | |
Primary | |
Store undiluted at -20°C. |
4D10 | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a | |
Affinity Purified |
For Research Use Only.