Learn More
Salmonella LPS Monoclonal Antibody (F62C), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17447
Additional Details : Weight : 0.01000kg
Description
The MA1-7447 antibody reacts with Salmonellae sp. common core Ag in bacterial and infected tissue samples. MA1-7447 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7447 antibody was raised against Salmonella common core antigen.
Salmonella LPS Monoclonal antibody specifically detects Salmonella LPS in Bacteria samples. It is validated for ELISA, Immunocytochemistry.Specifications
Salmonella LPS | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG2b |
ELISA, Immunocytochemistry | |
F62C | |
Unconjugated | |
Salmonellae | |
Common core antigen. | |
100 μg | |
Primary | |
4° C | |
Liquid |