missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSV Monoclonal Antibody (9C5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA120009
Additional Details : Weight : 0.01000kg
Description
MA1-20009 detects respiratory syncytial virus in virus samples.
RSV Monoclonal antibody specifically detects RSV in Virus samples. It is validated for ELISA.Specifications
RSV | |
Monoclonal | |
5.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG2b |
ELISA | |
9C5 | |
Unconjugated | |
human respiratory syncytial virus; RSV | |
Purified, full length, native RSV protein (long strain). | |
100 μg | |
Primary | |
4° C, do not freeze | |
Liquid |