missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB11FIP5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531790
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431. Predicted reactivity: Human (99%), Mouse (91%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RAB11FIP5 Polyclonal antibody specifically detects RAB11FIP5 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
RAB11FIP5 | |
Polyclonal | |
Unconjugated | |
RAB11FIP5 | |
9130206P09Rik; C75969; D6Ertd32e; GAF1; Gaf-1; Gamma-SNAP-associated factor 1; KIAA0857; Phosphoprotein pp75; pp75; RAB11 family interacting protein 5; RAB11 family interacting protein 5 (class I); rab11 family-interacting protein 5; Rab11fip5; Rab11-FIP5; rab11-interacting protein Rip11; Rip11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26056, 52055 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.75 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8R361, Q9BXF6 | |
RAB11FIP5 | |
Recombinant protein fragment corresponding to a region within amino acids 112 and 338 of Human RAB11-FIP5 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |