missing translation for 'onlineSavingsMsg'
Learn More

PRK1 Mouse, Unlabeled, Clone: 49, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610686

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15815589

  • 274.00€ / 1 set


Estimated Shipment: 12-10-2021
Add to basket

Description

Description

Members of the Protein Kinase C (PKC) family of homologous serine/threonine protein kinases are involved in a number of processes such as cell growth, cell differentiation, and cytokine secretion. PKCs are activated by Ca2+, phospholipids, diacylglycerol, phorbol esters, and proteolysis. PRK1 (PKC-Related Kinase 1, also named PKN) was originally identified in human hippocampus as a novel protein kinase with sequence homology to PKC. PRK1 contains 942 amino acids with an apparent molecular weight of 120kDa. Although activated by limited proteolysis, PRK1 is not activated by Ca2+/diacylglycerol or phorbol esters. However, PRK1 is activated by phospholipids and arachidonic acid. PRK1 may regulate cytoskeletal changes since it binds to Rho-GTP and becomes phosphorylated in vivo, coincidentally with the formation of focal adhesions and stress fibers.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

PRK1
49
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
PKN
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Videos
SDS
Documents
Certificates

For Research Use Only.