missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLUNC Rabbit anti-Bovine, Canine, Goat, Guinea Pig, Human, Mouse, Rabbit, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542187
Additional Details : Weight : 0.00010kg
Description
PLUNC is the human homolog of murine plunc and is specifically expressed in the upper airways and nasopharyngeal regions. The exact biological function of this protein is not known, however, it has been suggested to be involved in inflammatory responses to irritants in the upper airways. It may also serve as a potential molecular marker for detection of micrometastasis in non-small-cell lung cancer.This gene is the human homolog of murine plunc, and like the mouse gene, is specifically expressed in the upper airways and nasopharyngeal regions. The exact biological function of this gene is not known, however, it has been suggested to be involved in inflammatory responses to irritants in the upper airways. It may also serve as a potential molecular marker for detection of micrometastasis in non-small-cell lung cancer. Multiple transcript variants resulting from alternative splicing in the 3′ UTR have been detected, but the full-length nature of only two is known.Specifications
| PLUNC | |
| Polyclonal | |
| Unconjugated | |
| BPIFA1 | |
| Liquid | |
| BPIFA1 | |
| Synthetic peptide directed towards the middle region of human PLUNC | |
| Affinity Chromatography | |
| RUO | |
| Antibody | |
| Polyclonal | |
| Bovine, Canine, Goat, Guinea Pig, Human, Mouse, Rabbit, Rat |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8SPU5, Q9NP55, P97361, Q8K4I4 | |
| BPI fold containing family A member 1, LUNX, NASG, SPLUNC1, SPURT, bA49G10.5, PLUNC, LPLUNC3 | |
| Rabbit | |
| IgG | |
| 100 μL | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Primary | |
| 100349066, 100715971, 102186779, 18843, 246238, 281989, 51297, 609982 |