missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI3-Kinase Mouse, Unlabeled, Clone: 4, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610046
Additional Details : Weight : 0.01000kg
Description
PI3-kinase phosphorylates the D-3 position of the inositol ring of phosphatidylinositol (PtdIns), PtdIns(4)P and PtdIns(4,5)P2 to produce the respective PI3-phosphorylated derivatives. PI3-kinase exists as a heterodimer of 85 kDa (p85) and 110 kDa (p110) subunits. The p85 subunit contains two SH2 domains and an SH3 domain. It associates with and serves as a substrate for activated growth factor receptor tyrosine kinases. p85 may serve as regulator of the catalytic subunit, p110, by acting as the link between PI3-kinase and the ligand-activated receptor. Two distinct forms of the p85 subunit have been described: 1) p85 α; , which binds tightly to the catalytic subunit, and 2) p85ß, a protein whose function is presently unknown. Both isoforms bind to activated receptors and serve as tyrosine kinase substrates.Specifications
PI3-Kinase | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human PI3-Kinase α subunit aa. 562 to 724 | |
150 μg | |
Cell Biology | |
Canine, Chicken, Human, Mouse, Rat | |
IgG2a |
Western Blot | |
4 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.