Learn More
Phospho-EIF2S1 (Ser51) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537800
Additional Details : Weight : 0.00910kg
Description
A suggested positive control for Western blot is Jurkat cells; suggested positive control for IHC is human breast and human lung carcinoma; suggested positive control for ICC/IF is Hela cells.
Phospho-EIF2S1 (Ser51) Polyclonal antibody specifically detects Phospho-EIF2S1 (Ser51) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospho-EIF2S1 (Ser51) | |
Polyclonal | |
Unconjugated | |
EIF2S1 | |
0910001O23Rik; 2410026C18Rik; 35kDa; EIF2; EIF-2; eIF2 alpha; eIF2 alpha subunit; EIF2A; eIF-2A; eIF2alpha; EIF-2alpha; eIF-2-alpha; EIF2S1; eif2s1b; Eukaryotic translation initiation factor 2 subunit 1; eukaryotic translation initiation factor 2 subunit 1b; eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha b; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; fb19d01; translation initiation factor subunit; wu:fb19d01 | |
Rabbit | |
Affinity chromatography | |
RUO | |
13665, 1965, 54318 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P05198, P68101, Q6ZWX6 | |
EIF2S1 | |
Peptide sequence around phosphorylation site of serine 51 (E-L-S(p)-R-R) derived from Human eIF2-alpha | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |