Learn More
Nucleolin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA316875
Additional Details : Weight : 1.24740kg
Description
Heat mediated antigen retrieval is recommended for IHC on paraffin embedded tissues. Suggested positive control: PD31 murine pre-B cells for immunofluorescence and crude PD31 nuclear extracts for Western Blot.
Nucleolin Polyclonal antibody specifically detects Nucleolin in Human, Mouse, Plant, Rat samples. It is validated for ChIP Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot.Specifications
Nucleolin | |
Polyclonal | |
Unconjugated | |
NCL | |
B530004O11Rik; C23; D0Nds28; D1Nds28; FLJ45706; FLJ59041; Ncl; Nuc; Nucl; nucl antibody; Nucleolin; nucleolin-like protein; Protein C23 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Plant, Rat | |
Antibody | |
IgG |
ChIP Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot | |
Conc. Not Determined | |
whole serum with 0.1% sodium azide | |
P09405, P13383, P19338 | |
NCL | |
Fusion protein containing amino acids 284-709 of human nucleolin. | |
RUO | |
17975, 25135, 4691 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |