missing translation for 'onlineSavingsMsg'
Learn More

NBS1 Mouse, Unlabeled, Clone: 34, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611870

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15826049

  • 261.00€ / 1 set


Estimated Shipment: 08-10-2021
Add to basket

Description

Description

Nijmegen Breakage Syndrome (NBS) is characterized by extreme radiation sensitivity and chromosomal instability. The NBS1 gene product, p95/nibrin/NBS1, forms a complex with Rad50 and Mre11. Cells deficient in this complex have problems with DNA double-strand break repair, cell cycle checkpoint control, and telomere length maintenance. NBS1 contains a forkhead-associated domain (FHA) and a breast cancer carboxy-terminal domain (BRCT) in the N-terminal region. Both of these domains have been found in DNA-damage responsive cell cycle checkpoint proteins. The complex containing NBS1, Rad50, and Mre11 possesses manganese-dependent single stranded DNA endonuclease and 3′ to 5′ exonuclease activities. In addition, NBS1 is required for DNA-damage dependent phosphorylation of Mre11. This phosphorylation may be required for proper nuclear localization of the NBS1-Rad50-Mre11 complex to sites of DNA double-strand breaks. NBS1 interacts directly with telomere repeat binding factor, TRF1, via its C-terminal region, and both NBS1 and Mre11 co-localize with TRF1 at promyelocytic leukemia nuclear bodies. Thus, the NBS1-Rad50-Mre11 complex may be important for both DNA damage repair, and telomere length maintenance.

Host Species: Mouse
Clone: 34
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human NBS1 aa. 620-732

Immunofluorescence, Western Blotting

Specifications

Specifications

NBS1
34
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Nijmegen Breakage Syndrome-1
Affinity Purified
Human NBS1 aa. 620-732
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Videos
SDS
Documents
Certificates

For Research Use Only.