missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Mouse IgG2a, κ, Brilliant Violet 786, Clone: G155-178, BD
Brand: BD Biosciences 563732
Additional Details : Weight : 0.02000kg
Description
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a, κ
Immunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
G155-178 | |
Brilliant Violet 786 | |
Mouse | |
Affinity Purified | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
TNP-keyhole limpet hemocyanin | |
50 μg | |
Isotype Control (Primary) | |
IgG2a κ |