missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G and Ly-6C Rat anti-Mouse, Biotin, Clone: RB6-8C5, BD
Rat Monoclonal Antibody
363.00€ - 600.00€
Specifications
Antigen | Ly-6G and Ly-6C |
---|---|
Clone | RB6-8C5 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15897008
|
BD Biosciences
553124 |
0.1 mg |
363.00€
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
15807018
|
BD Biosciences
553125 |
0.5 mg |
600.00€
Each |
Estimated Shipment: 15-04-2024
Log in to see stock. |
|||||
Description
The RB6-8C5 antibody reacts with a common epitope on Ly-6G and Ly-6C, previously known as the myeloid differentiation antigen Gr-1. In the bone marrow, the level of antigen expression is directly correlated with granulocyte differentiation and maturation.
Host Species: Rat
Clone: RB6-8C5
Isotype: IgG2b, κ
Species Reactivity: Mouse
Cell Separation, Flow Cytometry
Specifications
Ly-6G and Ly-6C | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified | |
Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1 |
RB6-8C5 | |
Flow Cytometry | |
Biotin | |
RUO | |
Mouse | |
Not Reported | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.