missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki-67 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA516446
Additional Details : Weight : 0.01000kg
Description
PA5-16446 targets Ki-67 in IHC (P) applications and shows reactivity with Rat and Human samples. The PA5-16446 immunogen is a synthetic peptide from the human Ki67 protein.
Ki-67 Polyclonal antibody specifically detects Ki-67 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Ki-67 | |
Polyclonal | |
Unconjugated | |
Mki67 | |
antigen identified by monoclonal antibody Ki 67; antigen identified by monoclonal antibody Ki-67; Antigen identified by monoclonal antibody Ki-67 homolog; Antigen KI-67; Antigen KI-67 homolog; antigen KI-67; proliferation marker protein Ki-67; antigen KI-67-like; cb31; D630048A14Rik; I79_022666; Ki67; Ki-67; KIA; LOW QUALITY PROTEIN: proliferation marker protein Ki-67; marker of proliferation Ki-67; MIB-; MIB-1; Mki67; PPP1R105; Proliferation marker protein Ki-67; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105; RP11-380J17.2; sb:cb31; si:ch211-250b22.7; unnamed protein product; wu:fa11g09; wu:fb57a07; wu:fi14e05 | |
Rabbit | |
Protein A | |
RUO | |
291234, 4288 | |
4° C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P46013 | |
Mki67 | |
A synthetic peptide from the human Ki67 protein | |
500 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |