missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITK Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA517957
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse based on sequence homology.
ITK Polyclonal antibody specifically detects ITK in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
ITK | |
Polyclonal | |
Unconjugated | |
ITK | |
EMT; homolog of mouse T-cell itk/tsk; IL2 inducible T cell kinase; IL2 inducible T-cell kinase; IL-2-inducible T cell kinase; IL-2-inducible T-cell kinase; IL2-inducible T-cell kinase; interleukin-2-inducible T cell kinase; interleukin-2-inducible T-cell kinase; ITK; Kinase EMT; kinase TLK; LPFS1; LYK; MGC126257; MGC126258; PSCTK2; T-cell-specific kinase; Tcsk; Tlk; Tsk; Tyrosine-protein kinase ITK/TSK; tyrosine-protein kinase LYK | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3702 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q08881 | |
ITK | |
Synthetic peptide sequence (RLLRQLAEIAESGL) corresponding to the C-terminus amino acids of ITK | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |