missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA514824
Additional Details : Weight : 0.01000kg
Description
PDX1 Polyclonal antibody specifically detects PDX1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
PDX1 | |
Polyclonal | |
Unconjugated | |
PDX1 | |
DLDBP; E3BP; Glucose-sensitive factor; GSF; Idx1; IDX-1; Insulin promoter factor 1; insulin promoter factor 1, homeodomain transcription factor; insulin promotor factor 1; insulin upstream factor 1; Ipf1; IPF-1; Islet/duodenum homeobox 1; islet/duodenum homeobox-1; IUF1; IUF-1; MODY4; PAGEN1; Pancreas/duodenum homeobox protein 1; pancreatic and duodenal homeobox 1; pancreatic and duodenal homeobox gene 1; pancreatic-duodenal homeobox factor 1; PDHX; PDX1; pdx-1; somatostatin transactivating factor 1; somatostatin transcription factor 1; somatostatin-transactivating factor 1; Stf1; STF-1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
3651 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P52945 | |
PDX1 | |
KLH conjugated synthetic peptide between 186-215 amino acids from the C-terminal region of human IPF | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |