Learn More
IL-2 Rat anti-Mouse, APC, Clone: JES6-5H4, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554429
Additional Details : Weight : 0.01000kg
Description
The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2). The immunogen used to generate the JES6-5H4 hybridoma was recombinant mouse IL-2. This is a neutralizing antibody.
Host Species: Rat
Clone: JES6-5H4
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: Recombinant mouse IL-2
Intracellular Staining
Specifications
IL-2 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant mouse IL-2 | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b |
Flow Cytometry | |
JES6-5H4 | |
APC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.