missing translation for 'onlineSavingsMsg'
Learn More

IL-10 Rat anti-Mouse, FITC, Clone: JES5-16E3, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 554466

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15807718

  • 243.00€ / Each


Estimated Shipment: 13-10-2021
Add to basket

Description

Description

The JES5-16E3 antibody reacts with mouse interleukin-10 (IL-10). The immunogen used to generate the JES5-16E3 hybridoma was recombinant mouse IL-10. This is a neutralizing antibody.

Host Species: Rat
Clone: JES5-16E3
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: Recombinant mouse IL-10

Intracellular Staining

Specifications

Specifications

IL-10
JES5-16E3
FITC
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2b
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
Interleukin-10; Il10; CSIF; Cytokine synthesis inhibitory factor
Affinity Purified
Recombinant mouse IL-10
0.1mg
Immunology
Primary
Murine
Videos
SDS
Documents
Certificates

For Research Use Only.