missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG3, κ Isotype Control; Clone: J606; BD
Brand: BD Biosciences 563273
Additional Details : Weight : 0.01000kg
Description
Host Species: Mouse
Clone J606
Isotype IgG3, κ
Flow Cytometry
Specifications
| Control, Flow Cytometry | |
| 0.2mg/mL | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV510 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV510 were removed. | |
| RUO | |
| Isotype Control | |
| Monoclonal |
| J606 | |
| BV510 | |
| Affinity Purified | |
| Mouse BALB/c IgG3, κ | |
| 50μg | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
| Isotype Control |