Learn More
IFN alpha Monoclonal Antibody (7N41), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen M710B
Additional Details : Weight : 0.80000kg
Description
M710B targets IFN alpha in ELISA applications and shows reactivity with Human samples. The M710B immunogen is recombinant human IFN alpha 2b. M710B detects IFN alpha which has a predicted molecular weight of approximately 19 kDa.
IFN alpha Monoclonal antibody specifically detects IFN alpha in Human samples. It is validated for ELISA.Specifications
IFN alpha | |
Monoclonal | |
0.5 mg/mL | |
PBS with 4% BSA and proprietary preservative | |
P01562, P01563 | |
IFNA1, IFNA2 | |
Recombinant human IFN alpha 2b | |
250 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
7N41 | |
Biotin | |
IFNA1 | |
alpha-2a interferon; Gm12597; ifa; Ifa1; Ifa2; IFL; IFN; IFN alpha; IFN alpha 1/13; IFN alpha 1b; IFN a; IFN1@; IFNA; IFNA@; IFNA1; IFNA13; Ifna15; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFNA3; IFNA6; Ifnaa; IFNAalphaIFN-alpha-1/13; IFNalpha; IFN-alpha; IFN-alpha 1b; IFN-alpha1; IFN-alpha-1; IFN-alpha-1/13; IFN-alpha-2; IFN-alpha-6; IFN-alphaA; IFN-alphaD; IFN-gamma; IFNa; INFA2; interferon; interferon alpha; interferon alpha 1; Interferon alpha 13 (IFNA13); interferon alpha 15; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family gene 1; interferon alpha family, gene 2; Interferon alpha leukocyte (IFL); interferon alpha protein; interferon alpha-1; Interferon alpha1 (IFNA1); interferon alpha-1 precursor; interferon alpha-1/13; Interferon alpha-1/13 (IFN-alpha-1/13); Interferon alpha-2; Interferon alpha-A; Interferon alpha-D; Interferon alpha-D (IFN alphaD); interferon type A1/A2; interferon type A3; interferon, alpha 1; | |
Mouse | |
Caprylic acid precipitation | |
RUO | |
3439, 3440 | |
-20°C | |
Liquid |