missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA Tag Monoclonal Antibody (12CA5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA112429
Additional Details : Weight : 1.24740kg
Description
HA Tag Monoclonal antibody specifically detects HA Tag in Tag samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot.Specifications
HA Tag | |
Monoclonal | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
Mouse | |
Protein A/G | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
12CA5 | |
Unconjugated | |
HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
9 amino acid peptide to influenza virus hemagglutinin. | |
100 μg | |
Primary | |
Antibody | |
IgG2b |