missing translation for 'onlineSavingsMsg'
Learn More

H-2Kb Mouse anti-Mouse, PE, Clone: AF6-88.5, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 553570

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15807288

  • 298.00€ / Each


Estimated Shipment: 06-10-2021
Add to basket

Description

Description

The AF6-88.5 antibody reacts with the H-2Kb MHC class I alloantigen. Reactivity with other haplotypes (e.g., d, f, j, k, p, q, r, s, u, v) has not been observed.

Host Species: Mouse
Clone: AF6-88.5
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse C57BL splenocytes

Flow Cytometry

Specifications

Specifications

H-2Kb
AF6-88.5
PE
Affinity Purified
Mouse C57BL splenocytes
0.1mg
Immunology
Primary
Murine
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
IgG2a, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Videos
SDS
Documents
Certificates

For Research Use Only.