Learn More
GST-π Mouse, Unlabeled, Clone: 3, BD
Mouse Monoclonal Antibody
333.00€ - 660.00€
Specifications
Antigen | GST-π |
---|---|
Clone | 3 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15825609
|
BD Biosciences
610718 |
50 μg |
333.00€
1 set |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15835609
|
BD Biosciences
610719 |
150 μg |
660.00€
1 set |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
Description
Glutathione-S-Transferases (GSTs) are a family of dimeric proteins that catalyze the S-conjugation of glutathione with many compounds for detoxification. The GSTs are categorized into four classes based on their biochemical and structural properties: α, μ, π, and θ. GST-π is of particular interest because it is over-expressed in many tumors, but is either absent or expressed at low levels in the corresponding normal tissues. This high expression of GST-π is associated with malignant transformation and correlates with a decrease in GST activity. Three different GST-π proteins have been isolated and designated as hGSTP1™A, hGSTP1™B, and hGSTP1™C. These isoforms are nearly identical, varying by only one to two amino acids.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
GST-π | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Glutathione-S-Transferase-π |
3 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Human GST-π aa. 5-210 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.