Learn More
Glucocorticoid Receptor Mouse anti-Human, Unlabeled, Clone: 41, BD
Mouse Monoclonal Antibody
343.00€ - 690.00€
Specifications
Antigen | Glucocorticoid Receptor |
---|---|
Clone | 41 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15815849
|
BD Biosciences
611226 |
50 μg |
343.00€
1 set |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
15825849
|
BD Biosciences
611227 |
150 μg |
690.00€
1 set |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
Steroid hormone receptors are hormone activated transcriptional regulators that influence genes required for embryonic development and adult homeostasis. One member of the steroid hormone family is the glucocorticoid receptor. It contains AF1 and AF2 transactivation domains, a DNA binding domain, and ligand binding domain. Ligand bound glucocorticoid receptors dimerize at specific palindromic sequences called glucocorticoid response elements (GREs) in the cis-regulatory region of target genes. Both AF1 and AF2 may be important for initiation or regulation of transcription by interacting with components of the initiation complex or other intermediary factors. In addition to transactivation, glucocorticoid receptors may also regulate transcription through transrepression of target genes. Although mechanisms of transrepression are not completely understood; DNA binding alone, DNA binding plus interaction with other transcription factors, or protein-protein interaction without DNA binding are mechanisms that have been implicated. Thus, glucocorticoid receptors function in the
regulation of specific genes that are essential for human development and homeostasis.
Host Species: Mouse
Clone: 41
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Glucocorticoid Receptor α aa. 176-289
Formula Weight [Chemical]: 94kDa
Immunofluorescence, Western Blotting
Specifications
Glucocorticoid Receptor | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
41 | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Human Glucocorticoid Receptor α aa. 176-289 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.