missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTSJ1 Rabbit anti-Bovine, Canine, Equine, Human, Mouse, Porcine, Rabbit, Rat, Yeast, Zebrafish, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543300
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the S-adenosylmethionine-binding protein family. It is a nucleolar protein and may be involved in the processing and modification of rRNA. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq].Specifications
| FTSJ1 | |
| Polyclonal | |
| Unconjugated | |
| FTSJ1 | |
| Liquid | |
| FTSJ1 | |
| Synthetic peptide directed towards the N-terminal of human FTSJ1 | |
| Affinity Chromatography | |
| RUO | |
| Antibody | |
| Polyclonal | |
| Bovine, Canine, Equine, Human, Mouse, Porcine, Rabbit, Rat, Yeast, Zebrafish |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UET6 | |
| FtsJ homolog 1 (E. coli), CDLIV, JM23, MRX44, MRX9, SPB1, TRM7 | |
| Rabbit | |
| IgG | |
| 100 μL | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Primary | |
| 100061804, 100353966, 100431100, 24140, 363450, 480902, 492512, 517675, 54632 |