missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXP3 Monoclonal Antibody (22510), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514664
Additional Details : Weight : 0.01000kg
Description
Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with mouse based on sequence homology.
FOXP3 Monoclonal antibody specifically detects FOXP3 in Human, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
FOXP3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide; pH 7.5 | |
Q9BZS1 | |
Foxp3 | |
Synthetic peptide derived from residues 400 to the C-terminus of Human FOXP3. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
22510 | |
Unconjugated | |
Foxp3 | |
AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
Mouse | |
IgG fraction | |
RUO | |
317382, 50943 | |
-20°C or -80°C if preferred | |
Liquid |