Learn More
EBF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541632
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: LPSNCSSSSG IFSFSPANMV SAVKQKSAFA PVVRPQTSPP PTCTSTNGNS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
EBF1 Polyclonal antibody specifically detects EBF1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
EBF1 | |
Polyclonal | |
Unconjugated | |
EBF1 | |
COE1; Collier; Collier, Olf and EBF transcription factor 1; early B cell factor 1; early B-cell factor; early B-cell factor (olfactory neuronal transcription factor 1); early B-cell factor 1; Ebf; EBF transcription factor 1; EBF1; O/E-1; OE-1; OLF1; Olf-1; Olfactory neuronal transcription factor; olfactory neuronal transcription factor 1; transcription factor COE1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1879 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UH73 | |
EBF1 | |
Synthetic peptide directed towards the C-terminal of human EBF1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |