Learn More
Diphtheria Toxin Polyclonal Antibody, Biotin, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA173139
Additional Details : Weight : 0.01000kg
Description
The PA1-73139 antibody reacts with Diphtheria toxin. PA1-73139 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73139 antibody was raised against purified diphtheria toxoid.
Diphtheria Toxin Polyclonal antibody specifically detects Diphtheria Toxin in Bacteria samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Diphtheria Toxin | |
Polyclonal | |
Biotin | |
DT | |
Purified diphtheria toxoid. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goat | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG |