missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOPA Decarboxylase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525450
Additional Details : Weight : 0.01000kg
Description
DOPA Decarboxylase Polyclonal antibody specifically detects DOPA Decarboxylase in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
DOPA Decarboxylase | |
Polyclonal | |
Unconjugated | |
DDC | |
AADC; aromatic L-amino acid decarboxylase; aromatic-L-amino acid decarboxylase; aromatic-L-amino-acid decarboxylase; DDC; DOPA decarboxylase; dopa decarboxylase (aromatic L-amino acid decarboxylase); HGNC:2719; L-Dopa decarboxylase | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
13195, 1644 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
O88533, P20711 | |
DDC | |
KLH conjugated synthetic peptide between 32-61 amino acids from the N-terminal region of human DDC | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |