missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAGLB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526331
Additional Details : Weight : 0.01000kg
Description
DAGLB Polyclonal antibody specifically detects DAGLB in Human, Mouse samples. It is validated for Flow Cytometry, Western Blot.Specifications
DAGLB | |
Polyclonal | |
Unconjugated | |
DAGLB | |
DAGLB; DAGLBETA; DGL-beta; diacylglycerol lipase beta; diacylglycerol lipase, beta; E330036I19Rik; KCCR13L; RGD1310193; sn1-specific diacylglycerol lipase beta | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
221955, 231871 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide | |
Q8NCG7, Q91WC9 | |
DAGLB | |
KLH conjugated synthetic peptide between 491-518 amino acids from the central region of human DAGLB | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |