missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSP5 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530904
Additional Details : Weight : 0.01000kg
Specifications
| NSP5 | |
| Polyclonal | |
| Unconjugated | |
| SPECC1 | |
| Liquid | |
| SPECC1 | |
| Recombit fragment corresponding to a region within amino acids 437 and 731 of Human Cytospin B | |
| Antigen affinity chromatography | |
| RUO | |
| Antibody | |
| Polyclonal | |
| Human |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q5M775 | |
| CYTSB, HCMOGT-1, HCMOGT1, NSP, NSP5, cytokinesis and spindle organization B, cytospin-B, nuclear structure protein 5, sperm antigen HCMOGT-1, sperm antigen with calponin-like and coiled coil domains 1, structure protein NSP5a3a, structure protein NSP5a3b, structure protein NSP5b3a, structure pr | |
| Rabbit | |
| IgG | |
| 100 μL | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Primary | |
| 92521 |