missing translation for 'onlineSavingsMsg'
Learn More

Cyclin B1 Mouse, Unlabeled, Clone: GNS-11, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 554178

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15807608

  • 262.00€ / Each


Estimated Shipment: 13-10-2021
Add to basket

Description

Description

During the cell cycle, most eukaryotic cells double in mass, replicate their DNA and then distribute identical copies of their genome to progeny cells during mitosis. An internal biochemical clock ensures that all the necessary events occur in proper sequence and at the appropriate time. Cell in M phase contain a dominant regulatory factor known as maturation promoting factor (MPF). MPF triggers a variety of enzymatic and ultrastructural changes that are necessary for cell division. In higher eukaryotes, these mitosis-specific alterations include disassembly of the nuclear envelope, packaging of the DNA into chromosomes and assembly of the mitotic spindle. Purified MPF is a regulator of a protein kinase cascade and is evolutionarily conserved in all eukaryotic cells ranging from yeast to man. MPF consists predominantly of two polypeptides, cyclin B1 and p34, and contains protein kinase activity itself. It is the major M-phase-specific histone H1 kinase, but also phosphorylates a variety of other substrates including lamins, nucleolin, RNA polymerase II, retinoblastoma protein, SV40 large T antigen, p53, and the oncogenes c-src and c-abl. Cyclin B1 migrates at a reduced molecular weight of 62kDa on SDS-PAGE.

Host Species: Mouse
Clone: GNS-11
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Recombinant Human Cyclin B1
Formula Weight [Chemical]: 62kDa

Bioimaging, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

Cyclin B1
GNS-11
Unconjugated
Affinity Purified
Recombinant Human Cyclin B1
0.1mg
Cell Biology
Primary
Human, Murine
Western Blot
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
IgG2a
RUO
Store undiluted at 4°C
Monoclonal
Videos
SDS
Documents
Certificates

For Research Use Only.