missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CESK1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528468
Additional Details : Weight : 0.01000kg
Description
Possible molecular chaperone; assists the folding of proteins upon ATP hydrolysis.Specifications
| CESK1 | |
| Polyclonal | |
| Unconjugated | |
| CCT8L2 | |
| Liquid | |
| CCT8L2 | |
| Recombit fragment corresponding to a region within amino acids 352 and 557 of Human CESK1 | |
| Antigen affinity chromatography | |
| RUO | |
| Antibody | |
| Polyclonal | |
| Human |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96SF2 | |
| CESK1, T-complex protein 1, putative T-complex protein 1 subunit theta-like 2 | |
| Rabbit | |
| IgG | |
| 100 μL | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Primary | |
| 150160 |