missing translation for 'onlineSavingsMsg'
Learn More

CD95 Mouse, Unlabeled, Clone: 13, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610197

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15825329

  • 267.00€ / 1 set


Estimated Shipment: 13-10-2021
Add to basket

Description

Description

CD95 is a member of a family of cell surface receptors that includes tumor necrosis factor receptor (TNF-R), nerve growth factor receptor (NGF-R), CD40, CD27, CD30, and 4-1BB. Both murine and human Fas genes have been cloned and reportedly share 60% similarity in their amino acid sequences. CD95 (Fas) is a cell surface apoptosis-signaling molecule that is widely expressed in sites such as thymus, liver, heart, and ovary. Abnormalities in the Fas gene correlate with autoimmune features in mice and with unusually high levels of lymphocyte apoptosis in HIV-infected humans. Genetic studies have localized the Fas gene near the lpr (lymphoproliferation disease) locus on mouse chromosome 19 and further characterization reportedly has revealed that lpr is a mutation affecting the function of the Fas gene.

Specifications

Specifications

CD95
13
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG2a, κ
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Fas, APO-1
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Videos
SDS
Documents
Certificates

For Research Use Only.