missing translation for 'onlineSavingsMsg'
Learn More

CD49b Hamster anti-Rat, Unlabeled, Clone: HA1/29, BD

Hamster Monoclonal Antibody

Brand:  BD Biosciences 559987

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15809858

  • 183.00€ / Each


Estimated Shipment: 07-10-2021
Add to basket

Description

Description

The Ha1/29 antibody reacts with intergrin α2 chain (CD49b or VLA-2 α chain), the 160-kDa transmembrane glycoprotein that non-covalently associates with the intergrin β1 subunit (CD29) to form the intergrin α2β1 complex known as VLA-2. VLA-2, a receptor for collagen and laminin, is expressed on activated lymphocytes, epithelial cells, and platelets. It has been reported that Ha1/29 antibody can block VLA-2 binding to collagen and laminin. Ha1/29 antibody also reacts with mouse CD49b.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

CD49b
Ha1/29
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Hamster
IgG2, λ
RUO
Store undiluted at 4°C
Monoclonal
Flow Cytometry
0.5mg/mL
Integrin α2 chain
Affinity Purified
Cultured Rat Glomerular Epithelial
0.1mg
Immunology
Primary
Murine, Rat
Videos
SDS
Documents
Certificates

For Research Use Only.