missing translation for 'onlineSavingsMsg'
Learn More

CD45 Rat anti-Mouse, PE, Clone: 30-F11, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 553081

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15806988

  • 142.00€ / Each


Estimated Shipment: 08-10-2021
Add to basket

Description

Description

The 30-F11 clone has been reported to react with all isoforms and both alloantigens of CD45, which is found on hematopoietic stem cells and all cells of hematopoietic origin, except erythrocytes. CD45 is a transmembrane glycoprotein which is expressed at high levels on the cell-surface, and its presence distinguishes leukocytes from non-hematopoietic cells. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family, where the intracellular carboxy-terminal region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated as A, B, and C, respectively). CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction and the CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific.

Specifications

Specifications

CD45
30-F11
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2b, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Ptprc; CD45R; LCA; Leukocyte common antigen; T200; B220; Ly-5; Lyt-4
Affinity Purified
Mouse Thymus or Spleen
0.1mg
Immunology
Primary
Murine
Videos
SDS
Documents
Certificates

For Research Use Only.