missing translation for 'onlineSavingsMsg'
Learn More

CD45 Mouse anti-Human, Brilliant Violet 421, Clone: HI30, BD

Mouse Monoclonal Antibody

327.00€ - 760.00€

Specifications

Antigen CD45
Clone HI30
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Violet 421
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15833179
View Documents Promotion Details
BD Biosciences
563880
25 Tests
327.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Add to basket
 
15823179
View Documents Promotion Details
BD Biosciences
563879
100 Tests
760.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-04-2024
to see stock.
Add to basket
 
Description

Description

The HI30 monoclonal antibody specifically binds to the 180, 190, 205, 220 kDa protein isoforms of CD45. CD45 is encoded by the PTPRC ( Protein tyrosine phosphatase receptor type C) gene. CD45, also known as the leukocyte common antigen (LCA), is a member of the protein tyrosine phosphatase (PTP) family. It is present on all human leukocytes including lymphocytes, monocytes, granulocytes, eosinophils, and thymocytes. CD45 is absent from circulating erythrocytes, platelets, or mature erythroid cells of bone marrow and non-hemopoietic tissues.

Specifications

Specifications

CD45
Flow Cytometry
Brilliant Violet 421
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5
HI30
Monoclonal
Mouse
Human
Human Peripheral Blood Leucocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.