missing translation for 'onlineSavingsMsg'
Learn More

CD29 Mouse anti-Human, Brilliant Violet 510, Clone: MAR4, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 563513

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15822849

  • 451.00€ / Each


Estimated Shipment: 13-10-2021
Add to basket

Description

Description

The MAR4 monoclonal antibody specifically binds to CD29. CD29 is a 130 kDa integrin β1 subunit that is expressed as a heterodimeric complex with one of six distinct α subunits, comprising the very late activation antigen (VLA) subfamily of adhesion receptors. The β1 subunit has a broad tissue distribution; it is expressed on lymphocytes, monocytes and weakly on granulocytes, but not on erythrocytes. These receptors are involved in a variety of cell-cell and cell-matrix interactions.

Specifications

Specifications

CD29
Mar4
ITGB1; Integrin beta-1; Integrin β1; FNRB; GPIIA; ITB1; MDF2; MSK12; VLA-4β
Affinity Purified
Purified Human α5β1 Fibronectin Receptor
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 510
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Primary
Baboon, Cynomolgus, Human, Rhesus Monkey
Videos
SDS
Documents
Certificates

For Research Use Only.