missing translation for 'onlineSavingsMsg'
Learn More

CD279 Mouse anti-Human, PE, Clone: MIH4, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 560908

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15820589

  • 221.00€ / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket

Description

Description

The MIH4 monoclonal antibody specifically binds to the program death 1 (PD-1) receptor CD279, a member of the Ig superfamily. CD279 is an immunoregulatory receptor expressed on activated T cells, B cells and myeloid cells. Mice deficient in CD279 show a breakdown of peripheral tolerance and manifest multiple autoimmune symptoms. It contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) in the cytoplasmic region. PD-L1 and PD-L2 are ligands of CD279 and are members of the B7 gene family. Interaction of CD279:PD-Ligands results in inhibition of T cell proliferation and cytokine secretion. Reports suggest that the B7/CTLA-4 pathway functions primarily to attenuate, limit, and/or terminate naοve T-cell activation in secondary lymphoid organs. The PD-ligand:CD279 pathway, on the other hand, may primarily attenuate, limit, and/or terminate T-, B-, and myeloid cell activation/effector function at sites of inflammation in the periphery.

Specifications

Specifications

CD279
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human PD-1 Transfected Cell Line
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
MIH4
PD-1; hPD-1; PD1; PDCD1; Programmed cell death 1; SLEB2
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.