Learn More
CD25 Mouse anti-Human, FITC, Clone: M-A251, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560990
Additional Details : Weight : 0.01000kg
Description
The M-A251 monoclonal antibody specifically binds to the 55 kDa type I transmembrane glycoprotein known as the low-affinity interleukin-2 receptor alpha chain subunit (IL-2Rα). CD25 is expressed on regulatory T cells and on activated lymphocytes (T and B) and monocytes. It associates with the IL-2Rβ/CD122 and the IL-2Rγ/CD132 receptor chains to form the high-affinity IL-2R complex. CD25 expression on T and B lymphocytes is upregulated by antigenic or mitogenic stimulation. Soluble CD25/IL-2Rα is produced as a consequence of lymphocyte stimulation and is found in biological fluids following inflammatory responses.
Specifications
CD25 | |
Monoclonal | |
FITC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
M-A251 | |
IL-2R; IL2RA; IL-2Rα; TCGFR; TAC antigen; p55 | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.