missing translation for 'onlineSavingsMsg'
Learn More

CD21 Mouse anti-Human, APC, Clone: B-ly4, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 561767

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 15821319

  • 206.00€ / Each


Estimated Shipment: 06-10-2021
Add to basket

Description

Description

The B-ly4 monoclonal antibody specifically binds to CD21, a 145 kDa glycosylated type I integral membrane protein. CD21 is a receptor for the C3d complement fragment and for Epstein-Barr virus (EBV). CD21 is expressed on mature B cells, follicular dendritic cells, and some epithelial cells. It is also weakly expressed on the subset of mature T cells and thymocytes. CD21 plays a role in B-cell activation and proliferation. It may also play a role in modulating the function of T cells in the immune response to infections by lymphotropic viruses. Recently, CD21 was found to be part of a large complex containing CD19, CD81, and possibly other molecules.

Specifications

Specifications

CD21
B-ly4
CR2; Complement receptor type 2; C3DR; EBV-R; Epstein-Barr virus receptor
Affinity Purified
IgG1, κ
RUO
Primary
Baboon, Cynomolgus, Human, Porcine, Rhesus Monkey
Flow Cytometry
APC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
25 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Videos
SDS
Documenti
Certificati

For Research Use Only.