missing translation for 'onlineSavingsMsg'
Learn More

CD19 Rat anti-Mouse, BB515, Clone: 1D3, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 564531

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15813709

  • 146.00€ / Each


Estimated Shipment: 12-10-2021
Add to basket

Description

Description

The 1D3 antibody reacts with CD19, a B lymphocyte-lineage differentiation antigen. CD19, a 95-kDa transmembrance glycoprotein, is a member of the immunoglobulin superfamily and is expressed throughout B-lymphocyte development from the pro-B cell through the mature B-cell stages. Terminally differentiated plasma cells do not express CD19. On the surface of mature B cells, the CD19 molecule associates with CD21 (CR-2) and CD81 (TAPA-1), and this multimolecular complex synergizes with surface immunoglobulin to promote cellular activation. Studies with CD19-deficient mice have suggested that the level of CD19 expression affects the generation and maturation of B cells in the bone marrow and periphery. B-1 lineage B cells, also known as CD5+ B cells, are drastically reduced or absent in CD19-deficient mice. Increased levels of CD19 expression correlate with increased frequencies of peritonal and splenic B-1 cells and reduced numbers of conventional B lymphocytes in the periphery. CD19 participates in B-lymphocyte development, B-cell activation, maturation of memory B cells and regulation of tolerance. CD19 has also been detected on peritoneal mast cells, co-localized with CD21/CD35, and it is proposed to play a role in complement-mediated mast-cell activation.

The antibody was conjugated to BD Horizon™ BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

Specifications

Specifications

CD19
1D3
BB515
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Cd19; CD19 antigen; B-lymphocyte antigen CD19
Affinity Purified
Mouse CD19 Transfected Cell Line
25μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Videos
SDS
Documents
Certificates

For Research Use Only.